missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM Kinase 1 Antibody (OTI3H6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71141B
This item is not returnable.
View return policy
Description
LIM Kinase 1 Monoclonal specifically detects LIM Kinase 1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| LIM Kinase 1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| LIMK1 | |
| Full length recombinant protein of human LIM Kinase 1 (NP_002305) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3H6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 | |
| Mouse | |
| Affinity Purified | |
| Breast Cancer, Cancer, Phospho Specific, Protein Kinase | |
| 3984.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction