missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM kinase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 573.00
Specifications
| Antigen | LIM kinase 2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229200
|
Novus Biologicals
NBP3-38148-20ul |
20 μL |
N/A
|
N/A | |||||
|
30228567
|
Novus Biologicals
NBP3-38148-100ul |
100 μL |
€ 573.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIM kinase 2 Polyclonal antibody specifically detects LIM kinase 2 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| LIM kinase 2 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Phospho Specific, Protein Kinase | |
| PBS (pH 7.3), 50% glycerol | |
| 3985 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.7.11.1, LIM domain kinase 2, LIMK-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human LIM kinase 2 (NP_001026971.1).,, Sequence:, MGSYLSVPAYFTSRDLFRCSECQDSLTNWYYEKDGKLYCPKDYWGKFGEFCHGCSLLMTGPFMVAGEFKYHPECFACMSCKVIIEDGDAYALVQHATLYCGKCHNEVVLAPMFERLSTESVQEQLPYSVTLISMPATTEGRRGFSVSVESACSNYATTVQVKEVNRMHISPNNRNAIHPGDRILEINGTPVRTLRVEEVEDAISQTSQTLQLLIEHDPVSQRLDQLRLEARLAPHMQNAG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title