missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIM1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5116485

Product Code. 17268983

  • € 508.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive controls: A431 cell lysate, zebrafish tissue lysate, rat brian tissue, mouse brian tissue, MCF-7.

This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of neural and lymphoid cells. A similar protein in mice is an essential regulator of the vertebrate head organizer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LIM1
Polyclonal
Unconjugated
Lhx1
hLim-1; Homeobox protein Lim-1; LHX1; LHX-1; LIM homeo box protein 1; LIM homeobox 1; LIM homeobox protein 1; LIM/homeobox protein Lhx1; Lim1; Lim-1; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx1; MGC126723; MGC138141; rlim; transcription factor; homeobox protein
Rabbit
Antigen affinity chromatography
RUO
16869, 257634, 30463, 3975
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
P48742, P63006, P63007, Q90476
Lhx1
Synthetic peptide within zebrafish LIM1 aa 144-193.
100 μL
Primary
Human, Mouse, Rat, Zebrafish
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.