Learn More
Invitrogen™ LIM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121153
Description
Positive test controls include: BT-474, HepG2, Jurkat, Mouse liver, Mouse eye, Rat liver. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: TDHWMQYRLS GSFAHQGLWR YCLGNKCYLQ TDSIGEPPGQ GPGRAWGKSR ADLGAQGHLY SRWRTLRLKE GKGATQAYWN ATRAFM.
This gene encodes an eye lens-specific protein found at the junctions of lens fiber cells, where it may contribute to cell junctional organization. It acts as a receptor for calmodulin, and may play an important role in both lens development and cataractogenesis. Mutations in this gene have been associated with cataract formation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| LIM2 | |
| Polyclonal | |
| Unconjugated | |
| LIM2 | |
| 19kDa; 4833403J20; CTRCT19; Lens fiber membrane intrinsic protein; lens intrinsic membrane protein 2; lens intrinsic membrane protein 2, 19kDa; Lim2; MP17; MP18; Mp19; MP20; To3; total opacity 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114903, 233187, 3982 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P54825, P55344, P56563 | |
| LIM2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-110 of human LIM2 (NP_085915.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.