Learn More
LIMA1 Monoclonal Antibody (OTI7H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808601S
This item is not returnable.
View return policy
Description
LIMA1 Monoclonal antibody specifically detects LIMA1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| LIMA1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6P9G9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 203523 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7H5 | |
| Unconjugated | |
| ZNF449 | |
| 1110021C24Rik; D15Ertd366e; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; eplin; LIM domain and actin binding 1; LIM domain and actin-binding protein 1; LIMA1; mEPLIN; PP624; SREBP3; sterol regulatory element binding protein 3 | |
| Full length human recombinant protein of human ZNF449 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |