missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIME Antibody (LIME-10), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-343AF647
This item is not returnable.
View return policy
Description
LIME Monoclonal antibody specifically detects LIME in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| LIME | |
| Monoclonal | |
| Alexa Fluor 647 | |
| dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067 | |
| COOH-terminal peptide comprising residues 281-296 of the human LIME conjugated to keyhole limpet hemocyanin. | |
| 0.1 mL | |
| Signal Transduction | |
| 54923 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LIME-10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction