missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIME Antibody (OTI5D3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71974AF647
This item is not returnable.
View return policy
Description
LIME Monoclonal specifically detects LIME in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LIME | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LIME1 | |
| Recombinant protein fragment corresponding to amino acids 30-295 of human LIME (NP_060276) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5D3 | |
| Western Blot, Immunohistochemistry | |
| dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 54923.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction