missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Limitin/IFN-zeta Antibody (RM0123-7D19), Alexa Fluor™ 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12426AF405
This item is not returnable.
View return policy
Description
Limitin/IFN-zeta Monoclonal specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Limitin/IFN-zeta | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Ifnz | |
| Mouse recombinant Limitin/IFN-zeta | |
| 0.1 mL | |
| 319146 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0123-7D19 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 6030405N23Rik, BGIF, Ifnz, interferon zeta, Lmtn | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction