missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIMP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA131401
This item is not returnable.
View return policy
Description
Product purified by whole antiserum. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. SR-BI and SR-BII (previously known as SR-BI.2) are the alternatively spliced products of a single gene. SR-BII differs from SR-BI in that the encoded c-terminal cytoplasmic domain is almost completely different. SR-BII binds HDLs and mediates selective uptake of HDL cholesteryl ester, but with an approximately 4-fold lower efficiency than SR-BI. Nuclease protection assays show SR-BII to be abundant in mouse tissues expressing SR-BI, with SR-BII expression found in liver, adrenal glands, and testes. Although the role of SR-BII is not completely clear, research suggests that it may be a functional HDL receptor. In addition, SR-BII mRNA results from the alternative splicing of SR-BI precursor transcripts with the SR-BII isoform mediating selective transfer of lipid between HDL and cells. The relative expression and functional activities of these two isoforms create a potential means of regulating selective lipid transfer between HDL and cells.
Specifications
| LIMP2 | |
| Polyclonal | |
| Unconjugated | |
| SCARB2 | |
| 85 kDa lysosomal membrane sialoglycoprotein; 85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2; 9330185J12Rik; AMRF; CD 36; CD36; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2 (lysosomal integral membrane protein II); CD36 antigen-like 2; CD36 molecule; Cd36l2; EPM4; HLGP85; I79_021583; LGP85; LIMP II; LIMP2; LIMP-2; LIMPII; Lysosome membrane protein 2; lysosome membrane protein II; MLGP85; Scarb2; Scavenger receptor class B member 2; scavenger receptor class B, member 2; sCD 36; sCD36; soluble CD 36; soluble CD36; SR-B2; SR-BII | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Bovine, Hamster, Human, Mustelid, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 0.1% sodium azide | |
| O35114, P27615, Q14108 | |
| SCARB2 | |
| A peptide containing residues from mouse SR-BII (between residues 400-478) plus an N-terminal cysteine was coupled to KLH. [UniProt# O35114]. | |
| RUO | |
| 100771629, 117106, 12492, 533177, 950 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction