Learn More
LIMS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-31713
 View more versions of this product
Product Code. 18708413
  • € 624.00 / 0.10mL
Description
LIMS2 Polyclonal specifically detects LIMS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Rabbit
0.1 mL
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.