missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LIN-28A Antibody (PCRP-LIN28A-1E2) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27034PCP
This item is not returnable.
View return policy
Description
LIN-28A Monoclonal antibody specifically detects LIN-28A in Human samples. It is validated for Flow Cytometry
Specifications
| LIN-28A | |
| Monoclonal | |
| PerCP | |
| CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1 | |
| Recombinant fragment (around aa134-187) of human LIN-28A protein | |
| 0.1 mL | |
| Epigenetics, Neuroscience, Stem Cell Markers, Stem Cells | |
| 79727 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| PCRP-LIN28A-1E2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction