missing translation for 'onlineSavingsMsg'
Learn More
Learn More
lin-54 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82002AF350
This item is not returnable.
View return policy
Description
lin-54 Polyclonal antibody specifically detects lin-54 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| lin-54 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 350 | |
| CXCDC1, JC8.6, MIP120, Protein LIN-54 | |
| lin-54 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human lin-54. The immunogen is located within the last 50 amino acids of lin-54. Amino Acid Squence: EQADKKGKSKAAAERM | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 132660 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction