missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIN28A Monoclonal Antibody (14E6-4E6), DyLight™ 650
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA1016D650

Product Code. 15352867

  • € 495.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA1-016-D650 has been successfully used in ICC/IF and flow cytometry applications on human and mouse samples. MA1-016-D650 can be used for immunofluorescence analysis of LIN28 in human iPSCs.

Lin28 is a transcription factor that was first identified through its key role in the pathway of developmental timing in C. elegans. The role of Lin28 in development suggested that it might be useful in the creation of stem cells that might be beneficial in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing Lin28 in addition to the transcription factors POU5F1, Sox2, and NANOG in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Lin28, Sox2, POU5F1 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LIN28A
Monoclonal
1 mg/mL
PBS with proprietary stabilizer and 0.02% sodium azide
Q8K3Y3, Q9H9Z2
LIN28A
Full-length human recombinant protein expressed in bacteria.
100 μL
Primary
Human, Mouse
Antibody
IgG2a
Flow Cytometry, Immunocytochemistry
14E6-4E6
DyLight 650
LIN28A
AL024421; CSDD1; ENSMUSG00000070700; FLJ12457; Gm10299; Lin28; LIN-28; lin-28 homolog A; lin-28 homolog A (C. elegans); Lin28a; lin-28A; Protein lin-28 homolog A; RGD1566408; RNA-binding protein LIN-28; sb:cb650; si:ch211-232d9.4; similar to RNA-binding protein LIN-28; testis expressed gene 17; testis-expressed protein 17; Tex17; ZCCHC1; zgc:55584; Zinc finger CCHC domain-containing protein 1; zinc finger, CCHC domain containing 1
Mouse
Protein A
RUO
79727, 83557
4°C, do not freeze
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.