missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIN28B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550609
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total LIN28B protein.
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.
Specifications
| LIN28B | |
| Polyclonal | |
| Unconjugated | |
| LIN28B | |
| 2810403D23Rik; CSDD2; D030047M17Rik; lin-28 homolog B; lin-28 homolog B (C. elegans); Lin-28.2; Lin28b; Lin-28B; Protein lin-28 homolog B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 380669, 389421 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q45KJ6, Q6ZN17 | |
| LIN28B | |
| Synthetic peptide corresponding to residues near the C terminal of human lin-28 homolog B (C. elegans). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction