missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN52 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06592PCP
This item is not returnable.
View return policy
Description
LIN52 Polyclonal antibody specifically detects LIN52 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LIN52 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| c14_5549, C14orf46, chromosome 14 open reading frame 46, lin-52 homolog (C. elegans), protein lin-52 homolog | |
| Produced in rabbits immunized with E. coli-derived Human LIN52 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 91750 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction