missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN7B Antibody (OTI1C9), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72131MFV610
This item is not returnable.
View return policy
Description
LIN7B Monoclonal antibody specifically detects LIN7B in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| LIN7B | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI1C9 | |
| Flow Cytometry | |
| hLin7B, lin-7 homolog B (C. elegans), LIN-7B, MALS-2protein lin-7 homolog B, MALS2veli-2, Mammalian lin-seven protein 2, Veli-2, VELI2hVeli2, Vertebrate lin-7 homolog 2 | |
| Full length human recombinant protein of human LIN7B (NP_071448) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 64130 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction