missing translation for 'onlineSavingsMsg'
Learn More

LIN7B Monoclonal Antibody (OTI1C1), TrueMAB™, OriGene

Product Code. p-200010549
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17534361 30 μL 30µL
17514361 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17534361 Supplier Origene Technologies Supplier No. TA501479S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LIN7B Monoclonal antibody specifically detects LIN7B in Human samples. It is validated for Western Blot

Specifications

Antigen LIN7B
Applications Western Blot
Classification Monoclonal
Clone OTI1C1
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MIOX
Gene Accession No. Q9UGB7
Gene Alias hLin7B; hVeli2; lin 7 homolog b; lin-7 homolog B; lin-7 homolog B (C. elegans); lin-7 homolog B, crumbs cell polarity complex component; lin-7-A; LIN7B; lin-7B; MALS2; MALS-2; Mammalian lin-seven protein 2; protein lin-7 homolog B; UNQ3116/PRO10200; Veli1a; Veli2; Veli-2; veli-2 protein; vertebrate homolog of C. elegans Lin-7 type 2; vertebrate lin-7 homolog 2
Host Species Mouse
Immunogen Full length human recombinant protein of human MIOX produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55586
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.