missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN7B Monoclonal Antibody (OTI1E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501555S
This item is not returnable.
View return policy
Description
LIN7B Monoclonal antibody specifically detects LIN7B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| LIN7B | |
| Monoclonal | |
| 0.83 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HAP6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 64130 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1E3 | |
| Unconjugated | |
| LIN7B | |
| hLin7B; hVeli2; lin 7 homolog b; lin-7 homolog B; lin-7 homolog B (C. elegans); lin-7 homolog B, crumbs cell polarity complex component; lin-7-A; LIN7B; lin-7B; MALS2; MALS-2; Mammalian lin-seven protein 2; protein lin-7 homolog B; UNQ3116/PRO10200; Veli1a; Veli2; Veli-2; veli-2 protein; vertebrate homolog of C. elegans Lin-7 type 2; vertebrate lin-7 homolog 2 | |
| Full length human recombinant protein of human LIN7B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction