Learn More
Linezolid, 98%
Product Code. p-7077847 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15502393 250 mg 250mg
15592383 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15502393 Supplier Thermo Scientific Acros Supplier No. 460592500
Add to basket
missing translation for 'validMainframeMsg'
Linezolid, Zyvox, CAS # 165800-03-3 is a synthetic oxazolidinone antibiotic. | CAS: 165800-03-3 | C16H20FN3O4 | 337.35 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Linezolid is an organoflourine compound and an acetamide
• Linezolid acts by inhibiting initiation complex formation by binding to the 50S ribosomal subunit
Applications
• Linezolid is effective against staphylococcal and enterococcal microbes
• It is most effective against gram-positive organisms, particularly methicillin, vancomycin and penicillin resistant ones
• Suitable for research on antibiotic resistance, cross-reactivity and drug mechanisms
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 165800-03-3
Molecular Formula C16H20FN3O4
Molecular Weight (g/mol) 337.35
InChI Key TYZROVQLWOKYKF-ZDUSSCGKSA-N
Synonym linezolid, zyvox, zyvoxid, s-n-3-3-fluoro-4-morpholinophenyl-2-oxooxazolidin-5-yl methyl acetamide, zyvoxam, linezolide, linezlid, zyvoxa, linezolid usan:inn, unii-isq9i6j12j
PubChem CID 441401
ChEBI CHEBI:63607
IUPAC Name N-[[(5S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide
SMILES CC(=O)NCC1CN(C(=O)O1)C2=CC(=C(C=C2)N3CCOCC3)F
Specifications
Melting Point 181.0°C to 183.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 337.35
Percent Purity 98%
Chemical Name or Material Linezolid
compliance-icons
Product Identifier
  • Linezolid
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.