missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LINGO4 Polyclonal Antibody
Description
Sequence homology: Cow: 100%; Dog: 79%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
LINGO4 is a protein coding gene.
Specifications
Specifications
| Antigen | LINGO4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | LINGO4 |
| Gene Accession No. | Q6UY18 |
| Gene Alias | A530050P17Rik; DAAT9248; LERN4; leucine rich repeat and Ig domain containing 4; leucine rich repeat neuronal 6D; leucine-rich repeat and immunoglobulin-like domain containing-NOGO receptor-interacting protein 4; Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 4; leucine-rich repeat neuronal protein 6D; Lingo4; Lrrn6d; PRO34002; RGD1562025; UNQ9248/PRO34002 |
| Gene Symbols | LINGO4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?