missing translation for 'onlineSavingsMsg'
Learn More

Linifanib, MedChemExpress

Product Code. 16454017
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16454017

Brand: Medchem Express HY50751

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Linifanib (ABT-869) is a potent and orally active multi-target inhibitor of VEGFR and PDGFR family with IC50s of 4, 3, 66, and 4 nM for KDR, FLT1, PDGFRβ, and FLT3, respectively. Linifanib shows prominent antitumor activity. Linifanib has much less activity against unrelated RTKs, soluble tyrosine kinases, or serine/threonine kinases. Linifanib is a specific miR-10b inhibitor that blocks miR-10b biogenesis.

Linifanib (ABT-869) is a potent and orally active multi-target inhibitor of VEGFR and PDGFR family with IC50s of 4, 3, 66, and 4 nM for KDR, FLT1, PDGFRβ, and FLT3, respectively. Linifanib shows prominent antitumor activity. Linifanib has much less activity against unrelated RTKs, soluble tyrosine kinases, or serine/threonine kinases. Linifanib is a specific miR-10b inhibitor that blocks miR-10b biogenesis.

Specifications

Chemical Name or Material Linifanib
CAS 796967-16-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H18FN5O
Quantity 50 mg
Synonym ABT-869 AL-39324
Solubility Information DMSO : 13.16 mg/mL (35.06 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C2=C3C(N)=NNC3=CC=C2)C=C1)NC4=C(C=CC(C)=C4)F
Molecular Weight (g/mol) 375.4
Formula Weight 375.4
Percent Purity 98.3%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.