Learn More
Thermo Scientific Chemicals Lipase AY30
Product Code. 10297600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
10297600 25 g 25g
1 options
Product Code. 10297600
Brand: Thermo Scientific Chemicals 294760250
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Color Beige
For Use With (Application) Laboratory chemicals
Content And Storage 4°C
Quantity 25 g
Product Type Lipase AY30
Form Solid