missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipase I Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00149998-B01P
This item is not returnable.
View return policy
Description
Lipase I Polyclonal antibody specifically detects Lipase I in Human samples. It is validated for Western Blot
Specifications
| Lipase I | |
| Polyclonal | |
| PBS (pH 7.4) | |
| Cancer/testis antigen 17, CT17membrane-associated phospholipase A1 beta, EC 3.1.1, EC 3.1.1.-, EC 3.1.1.3, EC 3.1.1.34, lipase, member I, LPD lipase, LPDLlipase member I, Membrane-associated phosphatidic acid-selective phospholipase A1-beta, mPA-PLA1 beta, PRED5 | |
| LIPI (AAI48692.1, 1 a.a. - 481 a.a.) full-length human protein. MLLKCLHNNLCQKYSAHAFQFSPRNVLWLLVVCLRSDNKRPCLEFSQLSVKDSFRDLFIPRIETILMMYTRNNLNCAEPLFEQNNSLNVNFNTQKKTVWLIHGYRPVGSIPLWLQNFVRILLNEEDMNVIVVDWSRGATTFIYNRAVKNTRKVAVSLSVHIKNLLKHGASLDNFHFIGVSLGAHISGFVGKIFHGQLGRITGLDPAGPRFSRKPPYSRLDYTDAKFVDVIHSDSNGLGIQEPLGHIDFYPNGGNKQPGCPKSIFSGIQFIKCNHQRAVHLFMASLETNCNFISFPCRSYKDYKTSLCVDCDCFKEKSCPRLGYQAKLFKGVLKERMEGRPLRTTVFLDTSGTYPFCTYYFVLSIIVPDKTMMDGSFSFKLLNQLGMIEEPRLYEKNKPFYKLQEVKILAQFYNDFVNISSIGLTYFQSSNLQCSTCTYKIQRLMLKSLTYPERPPLCRYNIVLKDREEVFLNPNTCTPKNT | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAI48692.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 149998 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction