missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIPH Polyclonal Antibody
GREENER_CHOICE

Product Code. 15888943
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15888943 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15888943 Supplier Invitrogen™ Supplier No. PA570538

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

This gene encodes a membrane-bound member of the mammalian triglyceride lipase family. It catalyzes the production of 2-acyl lysophosphatidic acid (LPA), which is a lipid mediator with diverse biological properties that include platelet aggregation, smooth muscle contraction, and stimulation of cell proliferation and motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPH
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LIPH
Gene Accession No. Q8WWY8
Gene Alias AH; ARWH2; C130037N08Rik; D16Wsu119e; H06; HYPT7; Lacrimal lipase; LAH2; lipase H; lipase member H; lipase, member H; LIPH; LPD lipase-related protein; lpd1; lpd2; LPDLR; Membrane-associated phosphatidic acid-selective phospholipase A1-alpha; membrane-bound phosphatidic acid-selective phospholipase A1; MPAPLA1; mPA-PLA1; mPA-PLA1 alpha; P3; phospholipase A(1); phospholipase A1 member B; PLA1B
Gene Symbols LIPH
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human LIPH (aa 259-308).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200879
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.