missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-31920
This item is not returnable.
View return policy
Description
Lipin 3 Polyclonal specifically detects Lipin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Lipin 3 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q9BQK8 | |
| LPIN3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FFVQELESDDEHVPPGLCTSPIPWGGLSGFPSDSQLGTASEPEGLVMAGTASTGRRKRRRRRKPKQKEDAVATDSSP | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, phosphatidate phosphatase LPIN3, SMP2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 64900 | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur