missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipin 3 Antibody (OTI1D7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72160B
This item is not returnable.
View return policy
Description
Lipin 3 Monoclonal specifically detects Lipin 3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Lipin 3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| LPIN3 | |
| Full length recombinant protein of human Lipin 3 (NP_075047) produced in HEK293T cells. | |
| 0.1 mL | |
| 64900.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1D7 | |
| Western Blot, Immunohistochemistry | |
| dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, phosphatidate phosphatase LPIN3, SMP2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction