missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipin 3 Antibody (OTI1D7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72160CL1
This item is not returnable.
View return policy
Description
Lipin 3 Monoclonal antibody specifically detects Lipin 3 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Lipin 3 | |
| Monoclonal | |
| CoraFluor 1 | |
| dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, phosphatidate phosphatase LPIN3, SMP2 | |
| Full length human recombinant protein of human LPIN3 (NP_075047) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI1D7 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 64900 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction