missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipocalin-2/NGAL Antibody (14), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41364C
This item is not returnable.
View return policy
Description
Lipocalin-2/NGAL Monoclonal antibody specifically detects Lipocalin-2/NGAL in Canine, Human (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA, Sandwich ELISA Detection
Specifications
| Lipocalin-2/NGAL | |
| Monoclonal | |
| DyLight 650 | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| Recombinant dog NGAL | |
| 100 μg | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
| 3934 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA, Sandwich ELISA Detection | |
| 14 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Canine, Human (Negative), Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction