missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipocalin-2/NGAL Antibody (32), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05182AF532
This item is not returnable.
View return policy
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Lipocalin-2/NGAL | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LCN2 | |
| Recombinant rat NGAL precursor adsorbed onto aluminum hydroxide gel | |
| 0.1 mL | |
| Primary | |
| NGAL | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 32 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| Mouse | |
| Protein A or G purified | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
| 3934.0 | |
| Rat | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction