missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipocalin-2/NGAL Mouse anti-Monkey, Clone: 56, Novus Biologicals™
Mouse Monoclonal Antibody
€ 226.00 - € 618.00
Specifications
| Antigen | Lipocalin-2/NGAL |
|---|---|
| Clone | 56 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694018
|
Novus Biologicals
NBP2-41366 |
100 μg |
€ 618.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18101708
|
Novus Biologicals
NBP2-41366-0.025MG |
0.025 mg |
€ 226.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Human, Monkey samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA.Specifications
| Lipocalin-2/NGAL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| Recombinant monkey NGAL | |
| Primary | |
| Store at 4°C in the dark. |
| 56 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Monkey | |
| 3934 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title