missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIPT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15764961 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15764961 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15764961 Supplier Invitrogen™ Supplier No. PA557064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TDGTFLSSLL KSPYQGIRSN ATASIPSLVK NLLEKDPTLT CEVLMNAVAT EYAAYHQIDN HIHLINPTDE TLFPGINSKA KELQTWEWIY GKTPK Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 77%.

The process of transferring lipoic acid to proteins is a two-step process. The first step is the activation of lipoic acid by lipoate-activating enzyme to form lipoyl-AMP. For the second step, LIPT1 transfers the lipoyl moiety to apoproteins.The process of transferring lipoic acid to proteins is a two-step process. The first step is the activation of lipoic acid by lipoate-activating enzyme to form lipoyl-AMP. For the second step, the protein encoded by this gene transfers the lipoyl moiety to apoproteins. Alternative splicing in the 5' UTR of this gene results in five transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LIPT1
Gene Accession No. Q9Y234
Gene Alias Lipoate biosynthesis protein; Lipoate-protein ligase; Lipoyl ligase; lipoyltransferase 1; lipoyltransferase 1, mitochondrial; LIPT1; LIPT1D
Gene Symbols LIPT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LIPT1. Recombinant protein control fragment (Product #RP-96120).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51601
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.