missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIPT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559119
This item is not returnable.
View return policy
Description
Immunogen sequence: CAIGVRCGRH ITSHGLALNC STDLTWFEHI VPCGLVGTGV TSLSKELQRH VTVEEVMPPF LVAFKEIYKC TLISEDS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
Catalyzes the transfer of endogenously produced octanoic acid from octanoyl-acyl-carrier-protein onto the lipoyl domains of lipoate-dependent enzymes. Lipoyl-ACP can also act as a substrate although octanoyl-ACP is likely to be the physiological substrate.
Specifications
| LIPT2 | |
| Polyclonal | |
| Unconjugated | |
| LIPT2 | |
| Lipoate-protein ligase B; lipoyl(octanoyl) transferase 2 (putative); lipoyl/octanoyl transferase; LIPT2; Octanoyl-[acyl-carrier-protein]-protein N-octanoyltransferase; putative lipoyltransferase 2, mitochondrial; putative octanoyltransferase, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 387787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NK58 | |
| LIPT2 | |
| Recombinant protein corresponding to Human LIPT2. Recombinant protein control fragment (Product #RP-97493). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction