missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIS1 Recombinant Rabbit Monoclonal Antibody (23GB2825), Invitrogen™
GREENER_CHOICE

Product Code. p-200088326
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30088741 100 μL 100µL
30088727 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30088741 Supplier Invitrogen™ Supplier No. MA553526

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

VAMP7 is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family, localizing to late endosomes and lysosomes. VAMP7 is thought to mediate the fusion of endosomes to their target lysosomes as well as other exocytosis events during phagocytosis and neuritogenesis. VAMP7 interacts with the VPS9 ankyrin repeat protein VARP, a protein that localizes to early endosomes and thought to regulate endosome dynamics. Together with CD82, VAMP7 can modulate the signaling of EGFR by regulating its endocytosis from the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIS1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2825
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PAFAH1B1
Gene Accession No. P43034, P63004, P63005
Gene Alias HAMAP-Rule:MF_03141}; Lis1; Lis-1; LIS-1 {ECO:0000255; LIS2; lissencephaly 1 protein; lissencephaly-1 protein; lissencephaly-1 protein {ECO:0000255; MDCR; MDS; Mdsh; MGC25297; Miller-Dieker syndrome chromosome region; MMS10-U; Ms10u; NudF; PAF acetylhydrolase 45 kDa subunit; PAF acetylhydrolase 45 kDa subunit {ECO:0000255; PAFAH; PAF-AH 45 kDa subunit; PAF-AH 45 kDa subunit {ECO:0000255; PAFAH alpha; PAF-AH alpha; PAFAH alpha {ECO:0000255; PAF-AH alpha {ECO:0000255; PAF-AH beta; Pafah1b1; Pafaha; platelet activating factor acetylhydrolase 1b regulatory subunit 1; platelet activating factor acetylhydrolase 45 kDa subunit brain isoform; platelet activating factor acetylhydrolase 45 kDa subunit brain isoform variant; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa); platelet-activating factor acetylhydrolase beta subunit (PAF-AH beta); platelet-activating factor acetylhydrolase IB subunit alpha; platelet-activating factor acetylhydrolase IB subunit alpha {ECO:0000255; platelet-activating factor acetylhydrolase, isoform 1b, beta1 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 1; platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD); platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa; platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa); RP23-194P5.2
Gene Symbols PAFAH1B1
Host Species Rabbit
Immunogen A synthesized peptide derived from human LIS1 (55-90AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18472, 5048, 83572
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.