Learn More
Thermo Scientific Chemicals Lisinopril dihydrate
Product Code. 15871042 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15871042
Brand: Thermo Scientific Chemicals 462610010
Request Bulk or Custom Format
CAS: 83915-83-7 | C21H35N3O7 | 441.53 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83915-83-7
Molecular Formula C21H35N3O7
Molecular Weight (g/mol) 441.53
InChI Key CZRQXSDBMCMPNJ-ZUIPZQNBSA-N
PubChem CID 22887897
Specifications
Quantity 1 g
Chemical Name or Material Lisinopril dihydrate