missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lisinopril Dihydrate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Lisinopril Dihydrate
Specifications
Specifications
| Chemical Name or Material | Lisinopril Dihydrate |
| CAS | 83915-83-7 |
| Analyte or Component Names | Lisinopril Dihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H31 N3 O5 . 2 H2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Lisinopril Dihydrate, N2-[(1S)-1-Carboxy-3-phenylpropyl]-L-lysyl-L-proline dihydrate, (S)-1-[N2-(1-Carboxy-3-phenylpropyl)-L-lysyl]-L-proline dihydrate |
| InChI Formula | InChI=1S/C21H31N3O5.2H2O/c22-13-5-4-9-16(19(25)24-14-6-10-18(24)21(28)29)23-17(20(26)27)12-11-15-7-2-1-3-8-15;;/h1-3,7-8,16-18,23H,4-6,9-14,22H2,(H,26,27)(H,28,29);2*1H2/t16-,17-,18-;;/m0../s1 |
| SMILES | O.O.NCCCC[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2CCC[C@H]2C(=O)O |
| IUPAC Name | (2S)-1-[(2S)-6-amino-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]hexanoyl]pyrrolidine-2-carboxylic acid;dihydrate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction