Learn More
Lithium aluminum hydride, 95% min, Thermo Scientific™
Reducing Agent For Pharmaceuticals, Perfumes, and Fine organic Chemicals; Source of Hydrogen
Brand: Thermo Scientific Alfa Aesar 013442.14
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 16853-85-3 | |
| 95% min. | |
| MFCD00011075 | |
| Decomposes in water. Soluble in ether,diethyl ether and terahydrofuran. | |
| [Li+].[AlH4-] | |
| 37.95 | |
| 37.95 | |
| Air and moisture sensitive | |
| 0.917 g/cm3 at 20°C |
| 25g | |
| AlH4Li | |
| lithium aluminum hydride, lithium aluminium hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
| OCZDCIYGECBNKL-UHFFFAOYSA-N | |
| lithium(1+) alumanuide | |
| 21226445 | |
| Odorless | |
| Light gray powder | |
| Lithium aluminum hydride |
Safety and Handling
- Lithium aluminum hydride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contain: Lithium aluminum hydride