missing translation for 'onlineSavingsMsg'
Learn More

Lithium salicylate, tech.

Product Code. 11348118
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11348118

Brand: Thermo Scientific Alfa Aesar 039356.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 552-38-5 | C7H5LiO3 | 144.054 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-38-5
Molecular Formula C7H5LiO3
Molecular Weight (g/mol) 144.054
MDL Number MFCD00045813
InChI Key PSBOOKLOXQFNPZ-UHFFFAOYSA-M
Synonym lithium salicylate, unii-93f1sp6qin, salicylic acid lithium salt, dilithium salicylate, 93f1sp6qin, lithium 2-hydroxybenzoate, lithium 1+ ion salicylate, lithiumsalicylate, lithiumsalicylat, salicylicacidlithiumsalt
PubChem CID 23663621
IUPAC Name lithium;2-hydroxybenzoate
SMILES [Li+].C1=CC=C(C(=C1)C(=O)[O-])O

Specifications

Melting Point (decomposition)
Odor Sweet
Quantity 100 g
Solubility Information Soluble in water
Formula Weight 144.05
Grade Technical
Physical Form Powder
Chemical Name or Material Lithium salicylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.