missing translation for 'onlineSavingsMsg'
Learn More

Lithium triethylborohydride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 18873390 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal Glass Bottle
Unit Size:
800mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
18873390 800 mL AcroSeal Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18873390 Supplier Thermo Scientific Supplier No. 176998000
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22560-16-3 | Molecular Formula: C6H16BLi | Molecular Weight: 105.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22560-16-3 , 109-99-9
Molecular Formula C6H16BLi
Molecular Weight (g/mol) 105.94
MDL Number MFCD00011703
InChI Key WCJAYABJWDIZAJ-UHFFFAOYSA-N
Synonym Lithium triethylhydroborate
IUPAC Name lithium(1+) triethylboranuide
SMILES [Li+].CC[BH-](CC)CC

Specifications

Color Colorless to Gray
Concentration 0.9 to 1.1M
Density 0.8920g/mL
Packaging AcroSeal Glass Bottle
Linear Formula LiB(C2H5)3H
Quantity 800 mL
Fieser 04,313; 06,348; 08,309; 09,286; 10,249; 11,304; 12,289; 17,172
Merck Index 15, 5600
Physical Form Cloudy Solution
Formula Weight 105.94
Chemical Name or Material Lithium triethylborohydride, 1M Solution in THF
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.