Learn More
LIV-1/Zip6 Antibody (ladiratuzumAb) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28849JF585
This item is not returnable.
View return policy
Description
LIV-1/Zip6 Monoclonal antibody specifically detects LIV-1/Zip6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| LIV-1/Zip6 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| zinc transporter ZIP6, LIV-1, solute carrier family 39 (zinc transporter), member 6 | |
| LIV-1/ SLC39A6 | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 25800 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| ladiratuzumAb | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |