missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Livin Antibody (88C570), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56548AF532
This item is not returnable.
View return policy
Description
Livin Monoclonal antibody specifically detects Livin in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Livin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
| Recombinant human livin protein was used as immunogen. | |
| 0.1 mL | |
| Apoptosis, Zinc Finger | |
| 79444 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| 88C570 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction