missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Livin Antibody (88C570), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56548F
This item is not returnable.
View return policy
Description
Livin Monoclonal specifically detects Livin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Livin | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| BIRC7 | |
| Recombinant human livin protein was used as immunogen. | |
| 0.1 mL | |
| Apoptosis, Zinc Finger | |
| 79444 | |
| Human | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| 88C570 | |
| Immunocytochemistry/Immunofluorescence | |
| baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The Livin antibody, clone 88C570 has been shown to recognize both Livin alpha (∼39 kDa) and Livin beta (∼37 kDa) isoforms (Nachmias et al, 2003). 2. The antibody also recognizes the 30 kDa (p30-Livin alpha) and 28 kDa (p28-Livin beta) apoptotic cleavage fragments of Livin alpha and beta, respectively (Nachmias et al, 2003). Apoptotic cleavage fragments of Livin have also been referred to as truncated livin (tLivin) (Nachmias et al, 2007). 3. The specificity of the antibody has been validated using Livin shRNA (Quiping et al, 2004). | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction