missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Livin Antibody (OTI1D12), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72206APC
This item is not returnable.
View return policy
Description
Livin Monoclonal specifically detects Livin in Human samples. It is validated for Flow Cytometry.
Specifications
| Livin | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| BIRC7 | |
| Full length recombinant protein of human Livin (NP_071444) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI1D12 | |
| Flow Cytometry | |
| baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Zinc Finger | |
| 79444.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction