missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Livin Antibody (OTI1D12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02177
This item is not returnable.
View return policy
Description
Livin Monoclonal specifically detects Livin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Livin | |
| Monoclonal | |
| 0.36 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50 | |
| BIRC7 | |
| Full length human recombinant protein of human BIRC7 (NP_071444) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI1D12 | |
| Unconjugated | |
| baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
| Mouse | |
| 32.8 kDa | |
| 0.1 mL | |
| Apoptosis, Zinc Finger | |
| 79444 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction