missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LKAAEAR1 Antibody (Z36P2C1*E7), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50477IR
This item is not returnable.
View return policy
Description
LKAAEAR1 Monoclonal specifically detects LKAAEAR1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| C20orf201 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LKAAEAR1 | |
| Peptide Sequence - AIRLELFLPP (immunogen peptide is identical in mouse, rat, sheep, bovine and equine) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| Z36P2C1*E7 | |
| Western Blot, ELISA, Immunohistochemistry | |
| bA299N6.3, chromosome 20 open reading frame 201, hypothetical protein LOC198437, LKAAEAR motif containing 1, LKAAEAR1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 198437 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction