missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LKB1 Monoclonal Antibody (206R24)

Mouse Monoclonal Antibody
Marca: Invitrogen™ AHO1392
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LKB1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
LKB1 (also known as STK11 - Serine/threonine-protein kinase STK11), a member of the serine/threonine kinase family, is a tumor suppressor. LKB1 physically associates with p53 and regulates specific p53 dependent apoptosis pathways. LKB1 is present in both the cytoplasm and nucleus of living cells and translocates to mitochondria during apoptosis. LKB1 functions as a master upstream protein kinase, regulating AMPK related kinases as well as AMPK. Mutations in LKB1 gene are associated with Peutz Jeghers syndrome.
Especificaciones
| LKB1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
| D4AE59, Q15831, Q9WTK7 | |
| STK11 | |
| Recombinant fragment of human LKB1 expressed in E. coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 206R24 | |
| Unconjugated | |
| STK11 | |
| AA408040; CAMK group; CAMKL family; hLKB1; liver kinase B1; Liver kinase B1 homolog; LKB 1; LKB1; LKB-1; LKB1(S); Par-4; PJS; polarization-related protein LKB1; R75140; renal carcinoma antigen NY-REN-19; serine/threonine kinase 11; serine/threonine-protein kinase 11; serine/threonine-protein kinase LKB1; serine/threonine-protein kinase STK11; STK11 | |
| Mouse | |
| Purified | |
| RUO | |
| 20869, 314621, 6794 | |
| -20°C | |
| Liquid |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?