Learn More
LL-37 Antibody (B-F57), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18122MFV610
Product Code. 18505815
  • € 486.00 / 0.10mL
Description
LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LL-37
18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37
0.1 mL
Primary
Western Blot, ELISA