Learn More
LL-37 Antibody (B-L47), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18121MFV500
Product Code. 18576936
  • € 439.00 / 0.10mL
Description
LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LL-37
50mM Sodium Borate
Innate Immunity
Store at 4C in the dark.
ELISA
RUO
820