Learn More
LL-37 Antibody (B-L47), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18121MFV500
This item is not returnable.
View return policy
Description
LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for ELISA
Specifications
| LL-37 | |
| 50mM Sodium Borate | |
| Innate Immunity | |
| Store at 4C in the dark. |
| ELISA | |
| RUO | |
| 820 | |
| Purified |