missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMAN1 Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502269
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.Specifikationer
| LMAN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49257, Q62902 | |
| LMAN1 | |
| Full length human recombit protein of human LMAN1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1B8 | |
| Unconjugated | |
| LMAN1 | |
| ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 116666, 3998, 476186 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering