Learn More
LMAN1 Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502269S
Encompass_Preferred
 View more versions of this product
Product Code. 17556441
  • € 242.00 / 30µL
Description
LMAN1 Monoclonal antibody specifically detects LMAN1 in Human samples. It is validated for Western Blot
Specifications
Specifications
LMAN1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9GZU7
Mouse
Affinity chromatography
RUO
58190
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1B8
Unconjugated
CTDSP1
2610020P13Rik; AI326273; AU043785; C730041J05; EGK_09725; endoplasmic reticulum-golgi intermediate compartment protein 53; ERGIC53; ERGIC-53; ER-Golgi intermediate compartment 53 kDa protein; F5F8D; FMFD1; Gp58; intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose binding 1; lectin, mannose-binding, 1; LMAN1; MCFD1; MR60; P58; Protein ERGIC-53
Full length human recombinant protein of human CTDSP1 produced in E.coli.