Learn More
LMAN2 Antibody (02), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05830PE
Product Code. 18616756
  • € 407.00 / 0.10mL
Description
LMAN2 Monoclonal antibody specifically detects LMAN2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PBS
Signal Transduction
Human
C5orf8, chromosome 5 open reading frame 8, Glycoprotein GP36b, GP36B, Lectin mannose-binding 2, lectin, mannose-binding 2, vesicular integral protein of 36 kDa, vesicular integral-membrane protein VIP36, VIP36
10960